Assay Details
Gene Symbol: Pak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053306.3 | NP_445758.2 | 602 | 83 | ||
GenBank mRNA | S80221.1 | - | 521 | 83 | ||
U19967.1 | - | 602 | 83 | |||
U35345.1 | - | 521 | 83 |
Gene Symbol: LOC100910732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751066.2 | XP_003751114.1 | 5 - 6 | 750 | 83 | |
XM_006248473.1 | XP_006248535.1 | 6 - 7 | 879 | 83 | ||
XM_008768778.1 | XP_008767000.1 | 5 - 6 | 2311 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
