Assay Details
Gene Symbol: Pak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053306.3 | NP_445758.2 | 900 | 138 | ||
GenBank mRNA | S80221.1 | - | 819 | 138 | ||
U19967.1 | - | 900 | 138 | |||
U35345.1 | - | 819 | 138 |
Gene Symbol: LOC100910732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751066.2 | XP_003751114.1 | 8 - 9 | 1048 | 138 | |
XM_006248473.1 | XP_006248535.1 | 9 - 10 | 1177 | 138 | ||
XM_008768778.1 | XP_008767000.1 | 8 - 9 | 2609 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cell cycle
protein phosphorylation
phosphorylation
peptidyl-serine phosphorylation
intracellular signal transduction
regulation of apoptotic process
negative regulation of apoptotic process
protein autophosphorylation
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of protein tyrosine kinase activity
positive regulation of extrinsic apoptotic signaling pathway
negative regulation of cysteine-type endopeptidase activity involved in execution phase of apoptosis
protein serine/threonine kinase activity
ATP binding
protein kinase binding
protein tyrosine kinase activator activity
identical protein binding
mitotic cell cycle
protein phosphorylation
phosphorylation
peptidyl-serine phosphorylation
intracellular signal transduction
regulation of apoptotic process
negative regulation of apoptotic process
protein autophosphorylation
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of protein tyrosine kinase activity
positive regulation of extrinsic apoptotic signaling pathway
negative regulation of cysteine-type endopeptidase activity involved in execution phase of apoptosis
protein serine/threonine kinase activity
ATP binding
protein kinase binding
protein tyrosine kinase activator activity
identical protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P689755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553768 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39402 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P689755 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P689755 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P689755 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P689755 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24678 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 33451 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56439 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68541 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 702436586 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10751683 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751675 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751682 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751684 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751679 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751680 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751686 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751676 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751681 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751677 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751674 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751685 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751678 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751673 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10751671 | - | - |
Rat Genome 230 2.0 Array | 1388345_at | - | - |
Rat Genome 230 2.0 Array | 1393184_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369650_at | - | - |
Rat Genome 230 2.0 Array | 1369650_PM_at | - | - |
Rat Genome 230 2.0 Array | 1396267_at | - | - |
Rat Genome 230 2.0 Array | 1388345_PM_at | - | - |
Rat Genome 230 2.0 Array | 1396267_PM_at | - | - |
Rat Genome 230 2.0 Array | 1393184_at | - | - |
Affymetrix HT Rat Focus Array | 1396267_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369650_at | - | - |
Affymetrix HT Rat Focus Array | 1369650_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1396267_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1100004199.rn.1 | TC1100000763.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100004200.rn.1 | TC1100000763.rn.1 | RTA TC |
Rat Expression Set 230 | 1369650_at | - | - |
Rat Expression Set 230 | 1369650_PM_at | - | - |
Rat Expression Set 230 | 1396267_PM_at | - | - |
Rat Expression Set 230 | 1396267_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751671 | - | - |
Rat Genome U34 Set | U35345_s_at | - | - |
Rat Genome U34 Set | U35345_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21975446 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372206 | - | - |
RatRef-12 Expression BeadChip | ILMN_58549 | - | - |
