Assay Details
Gene Symbol: Asah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053407.3 | NP_445859.2 | 3 - 4 | 247 | 90 | |
GenBank mRNA | AF214647.1 | - | 3 - 4 | 227 | 90 | |
BC061540.1 | - | 3 - 4 | 250 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
