Assay Details
Gene Symbol: Kif3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053486.1 | NP_445938.1 | 5 - 6 | 2087 | 87 | |
GenBank mRNA | AF083330.1 | - | 5 - 6 | 2057 | 87 | |
AJ223599.1 | - | 5 - 6 | 2087 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P12822 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114785 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99450 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177784 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P12822 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P12822 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P12822 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12822 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702151137 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11970 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 41930 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46701 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 702151137 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10883318 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10883313 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10883315 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10883317 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10883320 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10883312 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10883319 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10883316 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10883314 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10883311 | - | - |
Rat Genome 230 2.0 Array | 1387657_at | - | - |
Rat Genome 230 2.0 Array | 1378891_PM_at | - | - |
Rat Genome 230 2.0 Array | 1378891_at | - | - |
Rat Genome 230 2.0 Array | 1387657_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369637_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369637_at | - | - |
Affymetrix HT Rat Focus Array | 1387657_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1378891_at | - | - |
Affymetrix HT Rat Focus Array | 1369637_at | - | - |
Affymetrix HT Rat Focus Array | 1378891_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387657_at | - | - |
Affymetrix HT Rat Focus Array | 1369637_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0600002173.rn.1 | TC0600000321.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600002179.rn.1 | TC0600000321.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600002174.rn.1 | TC0600000321.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600002170.rn.1 | TC0600000321.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600002171.rn.1 | TC0600000321.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600002175.rn.1 | TC0600000321.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600002178.rn.1 | TC0600000321.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600002180.rn.1 | TC0600000321.rn.1 | RTA TC |
Rat Expression Set 230 | 1387657_at | - | - |
Rat Expression Set 230 | 1369637_at | - | - |
Rat Expression Set 230 | 1387657_PM_at | - | - |
Rat Expression Set 230 | 1369637_PM_at | - | - |
Rat Expression Set 230 | 1378891_PM_at | - | - |
Rat Expression Set 230 | 1378891_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10883311 | - | - |
Rat Genome U34 Set | AF083330_at | - | - |
Rat Genome U34 Set | rc_AI013107_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21579678 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356741 | - | - |
RatRef-12 Expression BeadChip | ILMN_63093 | - | - |
