Assay Details
Gene Symbol: Tinagl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053582.2 | NP_446034.1 | 2 - 3 | 375 | 74 | |
XM_006238977.2 | XP_006239039.1 | 3 - 4 | 467 | 74 | ||
XM_006238978.2 | XP_006239040.1 | 3 - 4 | 464 | 74 | ||
GenBank mRNA | AB050717.1 | - | 2 - 3 | 376 | 74 | |
BC097299.1 | - | 3 - 4 | 434 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
