Assay Details
Gene Symbol: Gosr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053584.2 | NP_446036.1 | 4 - 5 | 349 | 83 | |
XM_006246914.1 | XP_006246976.1 | 4 - 5 | 417 | 83 | ||
XM_008768132.1 | XP_008766354.1 | 3 - 4 | 220 | 83 | ||
GenBank mRNA | BC126068.1 | - | 4 - 5 | 353 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
