Assay Details
Gene Symbol: Pdlim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053650.1 | NP_446102.1 | 4 - 5 | 477 | 68 | |
GenBank mRNA | AF002281.1 | - | 4 - 5 | 495 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P028895 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P447158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31014 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117631 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P447158 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P028895 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P028895 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P447158 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P447158 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P028895 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P447158 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P149321 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700292279 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 26270 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11986 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 27459 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700292279 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10791748 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791745 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791741 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791743 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791744 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791747 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791749 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791742 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10791740 | - | - |
Rat Genome 230 2.0 Array | 1370291_at | - | - |
Rat Genome 230 2.0 Array | 1370291_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370291_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370291_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1600012438.rn.1 | TC1600001659.rn.1 | RTA TC |
Rat Expression Set 230 | 1370291_at | - | - |
Rat Expression Set 230 | 1370291_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791740 | - | - |
Rat Genome U34 Set | AF002281_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22274586 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352755 | - | - |
RatRef-12 Expression BeadChip | ILMN_60040 | - | - |
