Assay Details
Gene Symbol: Vegfc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053653.1 | NP_446105.1 | 4 - 5 | 778 | 115 | |
GenBank mRNA | AF010302.1 | - | 2 - 3 | 235 | 115 | |
AY032729.1 | - | 4 - 5 | 778 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
