Assay Details
Gene Symbol: Cyp27b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053763.1 | NP_446215.1 | 1 - 2 | 194 | 70 | |
XM_008765374.1 | XP_008763596.1 | 2 - 3 | 2066 | 70 | ||
GenBank mRNA | AB001992.1 | - | 1 - 2 | 194 | 70 |
Gene Symbol: LOC100909462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773774.1 | XP_008771996.1 | 954 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
