Assay Details
Gene Symbol: Il2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053836.1 | NP_446288.1 | 2 - 3 | 220 | 84 | |
GenBank mRNA | M22899.1 | - | 2 - 3 | 220 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
