Assay Details
Gene Symbol: Kcnh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053937.1 | NP_446389.1 | 9 - 10 | 2182 | 125 | |
XM_006247556.2 | XP_006247618.1 | 9 - 10 | 2210 | 125 | ||
GenBank mRNA | AF016192.1 | - | 9 - 10 | 2182 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
