Assay Details
Gene Symbol: Cand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054004.2 | NP_446456.1 | 4 - 5 | 845 | 107 | |
GenBank mRNA | D87671.1 | - | 4 - 5 | 847 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
SCF complex assembly
protein ubiquitination
cell differentiation
negative regulation of catalytic activity
positive regulation of transcription, DNA-templated
positive regulation of RNA polymerase II transcription preinitiation complex assembly
TBP-class protein binding
transcription, DNA-templated
SCF complex assembly
protein ubiquitination
cell differentiation
negative regulation of catalytic activity
positive regulation of transcription, DNA-templated
positive regulation of RNA polymerase II transcription preinitiation complex assembly
TBP-class protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P13018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166021 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P13018 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P13018 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P13018 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13018 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59191 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88827 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20107 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10902643 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902642 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902638 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902636 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902635 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902631 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902639 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902637 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902645 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902640 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902644 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902632 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902633 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902641 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902634 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10902630 | - | - |
Rat Genome 230 2.0 Array | 1367972_PM_at | - | - |
Rat Genome 230 2.0 Array | 1367972_at | - | - |
Affymetrix HT Rat Focus Array | 1367972_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1367972_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700021729.rn.1 | TC0700002795.rn.1 | RTA TC |
Rat Expression Set 230 | 1367972_at | - | - |
Rat Expression Set 230 | 1367972_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902630 | - | - |
Rat Genome U34 Set | rc_AI230395_at | - | - |
Rat Genome U34 Set | E12829cds_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20827656 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1375217 | - | - |
RatRef-12 Expression BeadChip | ILMN_66995 | - | - |
