Assay Details
Gene Symbol: Cdk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080885.1 | NP_543161.1 | 1 - 2 | 129 | 89 | |
GenBank mRNA | L02121.1 | - | 1 - 2 | 129 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the L02121.1, NM_080885.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
