Assay Details
Gene Symbol: Cry2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133405.2 | NP_596896.2 | 5 - 6 | 740 | 64 | |
GenBank mRNA | AY033877.1 | - | 5 - 6 | 750 | 64 | |
BC166416.1 | - | 5 - 6 | 740 | 64 | ||
FQ211347.1 | - | 5 - 6 | 750 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
photoreactive repair
DNA repair
transcription, DNA-templated
protein import into nucleus
signal transduction
circadian rhythm
protein-chromophore linkage
lipid storage
negative regulation of phosphoprotein phosphatase activity
response to insulin
circadian regulation of gene expression
glucose homeostasis
regulation of circadian rhythm
negative regulation of circadian rhythm
entrainment of circadian clock by photoperiod
negative regulation of transcription, DNA-templated
regulation of sodium-dependent phosphate transport
negative regulation of glucocorticoid receptor signaling pathway
negative regulation of glucocorticoid secretion
transcription cis-regulatory region binding
DNA binding
damaged DNA binding
single-stranded DNA binding
deoxyribodipyrimidine photo-lyase activity
DNA photolyase activity
DNA (6-4) photolyase activity
photoreceptor activity
kinase binding
protein kinase binding
phosphatase binding
ubiquitin binding
FAD binding
negative regulation of transcription by RNA polymerase II
photoreactive repair
DNA repair
transcription, DNA-templated
protein import into nucleus
signal transduction
circadian rhythm
protein-chromophore linkage
lipid storage
negative regulation of phosphoprotein phosphatase activity
response to insulin
circadian regulation of gene expression
glucose homeostasis
regulation of circadian rhythm
negative regulation of circadian rhythm
entrainment of circadian clock by photoperiod
negative regulation of transcription, DNA-templated
regulation of sodium-dependent phosphate transport
negative regulation of glucocorticoid receptor signaling pathway
negative regulation of glucocorticoid secretion
transcription cis-regulatory region binding
DNA binding
damaged DNA binding
single-stranded DNA binding
deoxyribodipyrimidine photo-lyase activity
DNA photolyase activity
DNA (6-4) photolyase activity
photoreceptor activity
kinase binding
protein kinase binding
phosphatase binding
ubiquitin binding
FAD binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P050964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508014 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87012 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P050964 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P050964 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P050964 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13175 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97382 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23936 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10847528 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847533 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847529 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847532 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847526 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847537 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847534 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847530 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847531 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847527 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847535 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847536 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10847525 | - | - |
Rat Genome 230 2.0 Array | 1369446_at | - | - |
Rat Genome 230 2.0 Array | 1372548_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369446_PM_at | - | - |
Rat Genome 230 2.0 Array | 1372548_at | - | - |
Affymetrix HT Rat Focus Array | 1369446_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372548_at | - | - |
Affymetrix HT Rat Focus Array | 1372548_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369446_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300028053.rn.1 | TC0300003680.rn.1 | RTA TC |
Rat Expression Set 230 | 1372548_at | - | - |
Rat Expression Set 230 | 1369446_PM_at | - | - |
Rat Expression Set 230 | 1369446_at | - | - |
Rat Expression Set 230 | 1372548_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847525 | - | - |
Rat Genome U34 Set | rc_AI059856_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21323316 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351530 | - | - |
RatRef-12 Expression BeadChip | ILMN_49887 | - | - |
