Assay Details
Gene Symbol: Slco6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133412.1 | NP_596903.1 | 7 - 8 | 1465 | 115 | |
XM_006245576.2 | XP_006245638.1 | 7 - 8 | 1535 | 115 | ||
GenBank mRNA | AF321415.1 | - | 7 - 8 | 1465 | 115 |
Gene Symbol: LOC102554121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245586.2 | XP_006245648.1 | 7 - 8 | 1536 | 115 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P13179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139001 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36014 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105629 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101367 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31147 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115599 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P13179 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P13179 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P13179 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13179 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 96052 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 28727 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88641 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 87811 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 54698 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84587 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 39577 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10930127 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930125 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930113 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930122 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930115 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930114 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930116 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930117 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930118 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930119 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930123 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930124 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930121 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930120 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930126 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10930112 | - | - |
Rat Genome 230 2.0 Array | 1387494_PM_at | - | - |
Rat Genome 230 2.0 Array | 1387494_at | - | - |
Affymetrix HT Rat Focus Array | 1387494_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387494_at | - | - |
Rat Expression Set 230 | 1387494_PM_at | - | - |
Rat Expression Set 230 | 1387494_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930112 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21975423 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373407 | - | - |
RatRef-12 Expression BeadChip | ILMN_60224 | - | - |
