Assay Details
Gene Symbol: Slc2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138827.1 | NP_620182.1 | 1 - 2 | 233 | 60 | |
GenBank mRNA | BC061873.1 | - | 1 - 2 | 161 | 60 | |
M13979.1 | - | 1 - 2 | 233 | 60 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC061873.1, M13979.1, NM_138827.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
