Assay Details
Gene Symbol: Rin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139038.1 | NP_620607.1 | 9 - 10 | 2083 | 87 | |
XM_008760073.1 | XP_008758295.1 | 2509 | 87 | |||
GenBank mRNA | U80076.1 | - | 9 - 10 | 2083 | 87 |
Gene Symbol: LOC100911794 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749037.3 | XP_003749085.1 | 9 - 10 | 2365 | 87 | |
XM_008760195.1 | XP_008758417.1 | 8 - 9 | 2509 | 87 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
