Assay Details
Gene Symbol: Il3ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248989.2 | XP_006249051.1 | 10 - 11 | 3529 | 80 | |
GenBank mRNA | AF030243.1 | - | 5 - 6 | 733 | 80 | |
BP475867.1 | - | 2 - 3 | 80 | |||
FQ214452.1 | - | 7 - 8 | 1107 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
