Assay Details
Gene Symbol: Tnfsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145681.1 | NP_663714.1 | 3 - 4 | 330 | 86 | |
GenBank mRNA | AY115578.1 | - | 3 - 4 | 330 | 86 | |
EF030546.1 | - | 3 - 4 | 321 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
