Assay Details
Gene Symbol: Rcan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153724.2 | NP_714946.2 | 1 - 2 | 150 | 65 | |
GenBank mRNA | AB075973.1 | - | 1 - 2 | 106 | 65 | |
BC072478.1 | - | 1 - 2 | 150 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
