Assay Details
Gene Symbol: Elavl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238389.2 | XP_006238451.1 | 1 - 2 | 559 | 90 | |
XM_006238391.2 | 1 - 2 | 559 | 90 | |||
GenBank mRNA | AB098713.1 | - | 1 - 2 | 483 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB098713.1, XM_006238389.2, XM_006238391.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
