Assay Details
Gene Symbol: Ston2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135874.1 | NP_001129346.1 | 4 - 5 | 2519 | 90 | |
XM_006240406.2 | XP_006240468.1 | 5 - 6 | 3016 | 90 | ||
XM_006240407.2 | XP_006240469.1 | 5 - 6 | 2754 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
