Assay Details
Gene Symbol: Ppme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191838.1 | NP_001178767.1 | 12 - 13 | 1220 | 64 | |
GenBank mRNA | BC099160.1 | - | 3 - 4 | 208 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
