Assay Details
Gene Symbol: Pklr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012624.3 | NP_036756.3 | 5 - 6 | 615 | 64 | |
XM_006232593.2 | XP_006232655.1 | 5 - 6 | 746 | 64 | ||
GenBank mRNA | BC161827.1 | - | 4 - 5 | 605 | 64 | |
M11709.1 | - | 5 - 6 | 616 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
