Assay Details
Gene Symbol: Slc2a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012751.1 | NP_036883.1 | 6 - 7 | 865 | 72 | |
XM_006246596.2 | XP_006246658.1 | 7 - 8 | 838 | 72 | ||
GenBank mRNA | BC085757.1 | - | 6 - 7 | 910 | 72 | |
D28561.1 | - | 6 - 7 | 865 | 72 | ||
FQ216450.1 | - | 7 - 8 | 804 | 72 | ||
FQ232657.1 | - | 6 - 7 | 924 | 72 | ||
FQ233419.1 | - | 6 - 7 | 900 | 72 | ||
J04524.1 | - | 6 - 7 | 847 | 72 | ||
M25482.1 | - | 6 - 7 | 830 | 72 | ||
X14771.1 | - | 6 - 7 | 878 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
