Assay Details
Gene Symbol: Gnai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013106.1 | NP_037238.1 | 8 - 9 | 1170 | 82 | |
GenBank mRNA | FQ222046.1 | - | 8 - 9 | 1233 | 82 | |
J03219.1 | - | 8 - 9 | 1179 | 82 | ||
M20713.1 | - | 8 - 9 | 1170 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
vesicle fusion
adenylate cyclase-modulating G protein-coupled receptor signaling pathway
regulation of heart contraction
metabolic process
negative regulation of secretion
cell division
GTPase activity
protein binding
GTP binding
protein domain specific binding
G-protein beta/gamma-subunit complex binding
G protein-coupled serotonin receptor binding
GTPase activating protein binding
metal ion binding
vesicle fusion
adenylate cyclase-modulating G protein-coupled receptor signaling pathway
regulation of heart contraction
metabolic process
negative regulation of secretion
cell division
GTPase activity
protein binding
GTP binding
protein domain specific binding
G-protein beta/gamma-subunit complex binding
G protein-coupled serotonin receptor binding
GTPase activating protein binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P056669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326001 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66934 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P056669 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P056669 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P056669 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1019639 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700529452 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 55262 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 93899 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 99047 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57573 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700529452 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10825964 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825963 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825971 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825972 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825970 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825968 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825967 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825965 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825966 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825969 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10825962 | - | - |
Rat Genome 230 2.0 Array | 1368030_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368030_at | - | - |
Rat Genome 230 2.0 Array | 1368029_at | - | - |
Rat Genome 230 2.0 Array | 1368029_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368030_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368030_at | - | - |
Affymetrix HT Rat Focus Array | 1368029_at | - | - |
Affymetrix HT Rat Focus Array | 1368029_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200030956.rn.1 | TC0200004459.rn.1 | RTA TC |
Rat Expression Set 230 | 1368029_PM_at | - | - |
Rat Expression Set 230 | 1368030_PM_at | - | - |
Rat Expression Set 230 | 1368030_at | - | - |
Rat Expression Set 230 | 1368029_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825962 | - | - |
Rat Genome U34 Set | rc_AI228247_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22309076 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364579 | - | - |
RatRef-12 Expression BeadChip | ILMN_66649 | - | - |
