Assay Details
Gene Symbol: Nt5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021576.2 | NP_067587.2 | 4 - 5 | 994 | 63 | |
GenBank mRNA | BC081806.1 | - | 4 - 5 | 1024 | 63 | |
BC098665.1 | - | 4 - 5 | 994 | 63 | ||
J05214.1 | - | 4 - 5 | 1344 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
