Assay Details
Gene Symbol: Ntrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021589.1 | NP_067600.1 | 8 - 9 | 1243 | 91 | |
GenBank mRNA | M85214.1 | - | 8 - 9 | 1243 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
