Assay Details
Gene Symbol: Mapk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021746.1 | NP_068514.1 | 4 - 5 | 612 | 104 | |
XM_008765781.1 | XP_008764003.1 | 4 - 5 | 341 | 104 | ||
GenBank mRNA | BC081950.1 | - | 4 - 5 | 591 | 104 | |
X96488.1 | - | 4 - 5 | 612 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
