Assay Details
Gene Symbol: Fgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024145.2 | NP_077059.2 | 3 - 4 | 465 | 84 | |
XM_006239078.2 | XP_006239140.1 | 2 - 3 | 365 | 84 | ||
XM_006239079.2 | XP_006239141.1 | 3 - 4 | 397 | 84 | ||
XM_006239080.2 | XP_006239142.1 | 3 - 4 | 397 | 84 | ||
XM_006239081.2 | XP_006239143.1 | 3 - 4 | 478 | 84 | ||
XM_006239082.2 | XP_006239144.1 | 2 - 3 | 409 | 84 | ||
GenBank mRNA | BC062025.1 | - | 3 - 4 | 465 | 84 | |
X57018.1 | - | 3 - 4 | 406 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
