Assay Details
Gene Symbol: Begain | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111115.1 | NP_001104585.1 | 3 - 4 | 277 | 57 | |
NM_024163.2 | NP_077077.1 | 3 - 4 | 297 | 57 | ||
XM_008764858.1 | XP_008763080.1 | 4 - 5 | 695 | 57 | ||
XM_008764859.1 | XP_008763081.1 | 3 - 4 | 605 | 57 | ||
XM_008764860.1 | XP_008763082.1 | 3 - 4 | 807 | 57 | ||
GenBank mRNA | AF064868.1 | - | 3 - 4 | 332 | 57 | |
AF064869.1 | - | 3 - 4 | 290 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
