Assay Details
Gene Symbol: Gk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024381.2 | NP_077357.2 | 3 - 4 | 358 | 127 | |
XM_006256987.2 | XP_006257049.1 | 3 - 4 | 392 | 127 | ||
XM_006256988.2 | XP_006257050.1 | 3 - 4 | 392 | 127 | ||
XM_008773274.1 | XP_008771496.1 | 3 - 4 | 373 | 127 | ||
XM_008773275.1 | XP_008771497.1 | 3 - 4 | 373 | 127 | ||
XM_008773276.1 | XP_008771498.1 | 3 - 4 | 373 | 127 | ||
XM_008773278.1 | XP_008771500.1 | 3 - 4 | 372 | 127 | ||
GenBank mRNA | D16102.1 | - | 3 - 4 | 361 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
