Assay Details
Gene Symbol: Aco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024398.2 | NP_077374.2 | 17 - 18 | 2214 | 74 | |
GenBank mRNA | AJ243266.1 | - | 17 - 18 | 2215 | 74 | |
BC061999.1 | - | 17 - 18 | 2214 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P158928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144589 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P158928 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P158928 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P158928 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P198349 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701612296 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 910 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73404 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 70251 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6816 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701612296 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10898020 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898004 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898009 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898014 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898018 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898008 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898013 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898011 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898005 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898007 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898016 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898019 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898012 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898010 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898015 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898021 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898017 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898006 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10898003 | - | - |
Rat Genome 230 2.0 Array | 1367589_PM_at | - | - |
Rat Genome 230 2.0 Array | 1367589_at | - | - |
Affymetrix HT Rat Focus Array | 1367589_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1367589_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700011507.rn.1 | TC0700001509.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700011512.rn.1 | TC0700001509.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700011508.rn.1 | TC0700001509.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700011510.rn.1 | TC0700001509.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700011509.rn.1 | TC0700001509.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700011511.rn.1 | TC0700001509.rn.1 | RTA TC |
Rat Expression Set 230 | 1367589_PM_at | - | - |
Rat Expression Set 230 | 1367589_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898003 | - | - |
Rat Genome U34 Set | rc_AI170777_g_at | - | - |
Rat Genome U34 Set | rc_AI170777_at | - | - |
Rat Genome U34 Set | rc_AI230919_at | - | - |
Rat Genome U34 Set | rc_AI235320_at | - | - |
Rat Genome U34 Set | rc_AI103956_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21496728 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352530 | - | - |
RatRef-12 Expression BeadChip | ILMN_48841 | - | - |
