Assay Details
Gene Symbol: Psmc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031149.1 | NP_112411.1 | 5 - 6 | 339 | 83 | |
GenBank mRNA | AB000491.1 | - | 5 - 6 | 339 | 83 | |
BC058462.1 | - | 5 - 6 | 335 | 83 | ||
D83521.1 | - | 5 - 6 | 357 | 83 | ||
FQ213263.1 | - | 5 - 6 | 356 | 83 | ||
FQ213735.1 | - | 5 - 6 | 331 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
