Assay Details
Gene Symbol: Psmd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031331.1 | NP_112621.1 | 8 - 9 | 1038 | 61 | |
XM_006232924.2 | XP_006232986.1 | 8 - 9 | 1034 | 61 | ||
GenBank mRNA | AB017188.1 | - | 8 - 9 | 1038 | 61 | |
AF175575.1 | - | 8 - 9 | 970 | 61 | ||
BC060559.1 | - | 8 - 9 | 979 | 61 | ||
FQ209672.1 | - | 7 - 8 | 831 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
