Assay Details
Gene Symbol: Ighmbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031586.1 | NP_113774.1 | 6 - 7 | 911 | 114 | |
GenBank mRNA | AF199411.1 | - | 6 - 7 | 911 | 114 | |
BC099790.1 | - | 6 - 7 | 1107 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
