Assay Details
Gene Symbol: Psmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031595.1 | NP_113783.1 | 11 - 12 | 1445 | 109 | |
GenBank mRNA | BC062019.1 | - | 11 - 12 | 1355 | 109 | |
D83522.1 | - | 11 - 12 | 1319 | 109 | ||
FQ225947.1 | - | 10 - 11 | 1104 | 109 | ||
FQ226273.1 | - | 10 - 11 | 1104 | 109 | ||
FQ233836.1 | - | 10 - 11 | 1104 | 109 | ||
U77918.1 | - | 11 - 12 | 1445 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
