Assay Details
Gene Symbol: Psmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031595.1 | NP_113783.1 | 7 - 8 | 967 | 82 | |
GenBank mRNA | BC062019.1 | - | 7 - 8 | 877 | 82 | |
D83522.1 | - | 7 - 8 | 841 | 82 | ||
FQ225567.1 | - | 6 - 7 | 626 | 82 | ||
FQ226273.1 | - | 6 - 7 | 626 | 82 | ||
FQ233836.1 | - | 6 - 7 | 626 | 82 | ||
U77918.1 | - | 7 - 8 | 967 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
