Assay Details
Gene Symbol: Nr1h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031626.1 | NP_113814.1 | 7 - 8 | 1128 | 60 | |
GenBank mRNA | BC157812.1 | - | 7 - 8 | 1107 | 60 | |
U14533.1 | - | 7 - 8 | 1128 | 60 | ||
U20389.1 | - | 7 - 8 | 1072 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
