Assay Details
Gene Symbol: Gosr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031685.2 | NP_113873.1 | 2 - 3 | 102 | 66 | |
GenBank mRNA | BC061994.1 | - | 2 - 3 | 102 | 66 | |
U91539.1 | - | 2 - 3 | 101 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
