Assay Details
Gene Symbol: Homer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031707.1 | NP_113895.1 | 3 - 4 | 591 | 72 | |
XM_006231775.2 | XP_006231837.1 | 3 - 4 | 1172 | 72 | ||
XM_008760635.1 | XP_008758857.1 | 3 - 4 | 1159 | 72 | ||
GenBank mRNA | AB003726.1 | - | 3 - 4 | 967 | 72 | |
AB007688.1 | - | 3 - 4 | 598 | 72 | ||
AB017140.1 | - | 3 - 4 | 591 | 72 | ||
AF030088.2 | - | 3 - 4 | 1502 | 72 | ||
AF093267.1 | - | 3 - 4 | 1036 | 72 | ||
AF093268.1 | - | 3 - 4 | 1028 | 72 | ||
AJ276327.1 | - | 3 - 4 | 311 | 72 | ||
AJ276328.1 | - | 3 - 4 | 311 | 72 | ||
U92079.1 | - | 3 - 4 | 863 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
