Assay Details
Gene Symbol: Homer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031707.1 | NP_113895.1 | 4 - 5 | 680 | 106 | |
XM_006231775.2 | XP_006231837.1 | 4 - 5 | 1261 | 106 | ||
XM_008760635.1 | XP_008758857.1 | 4 - 5 | 1248 | 106 | ||
GenBank mRNA | AB003726.1 | - | 4 - 5 | 1056 | 106 | |
AB007688.1 | - | 4 - 5 | 687 | 106 | ||
AB017140.1 | - | 4 - 5 | 680 | 106 | ||
AF030088.2 | - | 4 - 5 | 1591 | 106 | ||
AF093267.1 | - | 4 - 5 | 1125 | 106 | ||
AF093268.1 | - | 4 - 5 | 1117 | 106 | ||
AJ276327.1 | - | 4 - 5 | 400 | 106 | ||
AJ276328.1 | - | 4 - 5 | 400 | 106 | ||
U92079.1 | - | 4 - 5 | 952 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
