Assay Details
Gene Symbol: Adrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031708.1 | NP_113896.1 | 7 - 8 | 930 | 90 | |
XM_006235822.1 | XP_006235884.1 | 7 - 8 | 1051 | 90 | ||
GenBank mRNA | AB032742.1 | - | 7 - 8 | 930 | 90 | |
BC061773.1 | - | 7 - 8 | 946 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
