Assay Details
Gene Symbol: Htra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031721.1 | NP_113909.1 | 6 - 7 | 1161 | 83 | |
GenBank mRNA | AF179370.1 | - | 6 - 7 | 1161 | 83 | |
BC081767.1 | - | 6 - 7 | 1186 | 83 | ||
FQ220294.1 | - | 3 - 4 | 252 | 83 | ||
FQ228889.1 | - | 5 - 6 | 613 | 83 | ||
FQ228942.1 | - | 5 - 6 | 639 | 83 | ||
FQ229223.1 | - | 1 - 2 | 52 | 83 | ||
FQ229948.1 | - | 3 - 4 | 310 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
