Assay Details
Gene Symbol: Slc2a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031741.1 | NP_113929.1 | 1 - 2 | 71 | 83 | |
GenBank mRNA | BC076378.1 | - | 1 - 2 | 56 | 83 | |
BC161843.1 | - | 1 - 2 | 58 | 83 | ||
D13871.1 | - | 1 - 2 | 71 | 83 | ||
L05195.1 | - | 1 - 2 | 43 | 83 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC076378.1, BC161843.1, D13871.1, L05195.1, NM_031741.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
