Assay Details
Gene Symbol: Bcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031752.2 | NP_113940.1 | 11 - 12 | 1467 | 81 | |
GenBank mRNA | AB035510.1 | - | 11 - 12 | 1464 | 81 | |
BC072479.1 | - | 11 - 12 | 1467 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
