Assay Details
Gene Symbol: Cpz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031766.1 | NP_113954.1 | 4 - 5 | 779 | 81 | |
XM_006251151.2 | XP_006251213.1 | 3 - 4 | 774 | 81 | ||
GenBank mRNA | AF017637.1 | - | 4 - 5 | 779 | 81 | |
AF017638.1 | - | 4 - 5 | 779 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
