Assay Details
Gene Symbol: Fcgrt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033351.2 | NP_203502.1 | 5 - 6 | 1197 | 65 | |
XM_006229043.1 | XP_006229105.1 | 6 - 7 | 1306 | 65 | ||
XM_008759343.1 | XP_008757565.1 | 6 - 7 | 1205 | 65 | ||
GenBank mRNA | BC061975.1 | - | 6 - 7 | 1149 | 65 | |
FQ229080.1 | - | 5 - 6 | 979 | 65 | ||
M35495.1 | - | 5 - 6 | 1199 | 65 | ||
X14323.1 | - | 5 - 6 | 1199 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
