Assay Details
Gene Symbol: Ppp1r9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053474.1 | NP_445926.1 | 3 - 4 | 2021 | 66 | |
XM_008768126.1 | XP_008766348.1 | 2 - 3 | 1529 | 66 | ||
GenBank mRNA | AF016252.1 | - | 3 - 4 | 2021 | 66 | |
BC160878.1 | - | 3 - 4 | 1703 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
