Assay Details
Gene Symbol: Herpud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053523.1 | NP_445975.1 | 7 - 8 | 1098 | 69 | |
XM_006255133.2 | XP_006255195.1 | 7 - 8 | 1411 | 69 | ||
GenBank mRNA | AB033771.1 | - | 7 - 8 | 1098 | 69 | |
BC061849.1 | - | 7 - 8 | 1120 | 69 | ||
FQ219047.1 | - | 2 - 3 | 243 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
